Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fas Ligand/TNFSF6, Clone: MFL3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327605MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432765

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Antigen Fas Ligand/TNFSF6
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 356
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Format Affinity Purified
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD95 Ligand.
Regulatory Status RUO
Cross Reactivity Mouse
Concentration 0.5mg/mL
Clone MFL3
Formulation Phosphate buffer pH 7.2
Quantity 0.5mg
Isotype IgG
Purification Method Affinity Purified