Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fas Ligand/TNFSF6, Clone: MFL3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327601

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Regulatory Status RUO
Clone MFL3
Conjugate Unlabeled
Antigen Fas Ligand/TNFSF6
Format Affinity Purified
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD95 Ligand.
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 356
Concentration 0.5mg/mL
Cross Reactivity Mouse
Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified