Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fas Ligand/TNFSF6, Clone: MFL3, Biotin, Novus Biologicals™

$197.00 - $725.00

Specifications

Antigen Fas Ligand/TNFSF6
Clone MFL3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD95 Ligand.
Gene ID (Entrez) 356
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14352005

 
novus biologicals canada
NBP143520005MG
0.05mg Each for $197.00

NBP14352001

 
novus biologicals canada
NBP14352001MG
0.1mg Each for $329.00

NBP1435205

 
novus biologicals canada
NBP14352005MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen Fas Ligand/TNFSF6
Clone MFL3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD95 Ligand.
Gene ID (Entrez) 356
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry