Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD84/SLAMF5, Clone: mCD84.7, APC, Novus Biologicals™

$128.00 - $321.00

Specifications

Antigen CD84/SLAMF5
Clone mCD84.7
Immunogen The immunogen for this antibody was CD84.
Gene ID (Entrez) 8832
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14302901

 
novus biologicals canada
NBP14302901MG
0.1mg Each for $321.00

NBP14302903

 
novus biologicals canada
NBP1430290025MG
0.025mg Each for $128.00
Description & Specifications

Specifications

Antigen CD84/SLAMF5
Clone mCD84.7
Immunogen The immunogen for this antibody was CD84.
Gene ID (Entrez) 8832
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG1
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry