Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD84/SLAMF5, Clone: mCD84.7, APC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14302901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14302901

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Antigen CD84/SLAMF5
Format Affinity Purified
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD84.
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8832
Clone mCD84.7
Conjugate APC
Formulation Aqueous buffer (contains stabilizer if necessary)
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal