Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD84/SLAMF5, Clone: mCD84.7, APC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1430290025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14302903

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Applications Flow Cytometry
Isotype IgG1
Quantity 0.025mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8832
Conjugate APC
Cross Reactivity Mouse
Antigen CD84/SLAMF5
Host Species Armenian Hamster
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Immunogen The immunogen for this antibody was CD84.
Clone mCD84.7