Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD81, Clone: Eat-2, Novus Biologicals™

$246.00 - $397.00

Specifications

Antigen CD81
Clone Eat-2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD81.
Gene ID (Entrez) 975
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14338305

 
novus biologicals canada
NBP143383005MG
0.05mg Each for $246.00

NBP14338301

 
novus biologicals canada
NBP14338301MG
0.1mg Each for $300.00

NBP1433835

 
novus biologicals canada
NBP14338305MG
0.5mg Each for $397.00
Description & Specifications

Specifications

Antigen CD81
Clone Eat-2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD81.
Gene ID (Entrez) 975
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting