Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD81, Clone: Eat-2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14338305MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433835

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.5mg
Primary or Secondary Primary
Cross Reactivity Mouse
Antigen CD81
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was CD81.
Isotype IgG
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 975
Clone Eat-2
Concentration 0.5mg/mL
Conjugate Unlabeled
Host Species Armenian Hamster
Purification Method Affinity Purified