Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD81, Clone: Eat-2, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14338301MG

 View all options for this product


For Research Use Only

Catalog No. NBP14338301


Description & Specifications


Antigen CD81
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Clone Eat-2
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD81.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 975