Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD81, Clone: Eat-2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143383005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14338305

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD81.
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 975
Concentration 0.5mg/mL
Cross Reactivity Mouse
Antigen CD81
Isotype IgG
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone Eat-2
Conjugate Unlabeled