Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD69, Clone: H1.2F3, Biotin, Novus Biologicals™

$171.00 - $418.00

Specifications

Antigen CD69
Clone H1.2F3
Concentration 0.2mg/mL
Immunogen Y245 mouse dendritic epidermal T cell line (1)
Gene ID (Entrez) 969
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14362205

 
novus biologicals canada
NBP143622005MG
0.05mg Each for $171.00

NBP14362201

 
novus biologicals canada
NBP14362201MG
0.1mg Each for $246.00

NBP1436225

 
novus biologicals canada
NBP14362205MG
0.5mg Each for $418.00
Description & Specifications

Specifications

Antigen CD69
Clone H1.2F3
Concentration 0.2mg/mL
Immunogen Y245 mouse dendritic epidermal T cell line (1)
Gene ID (Entrez) 969
Formulation Phosphate buffer pH 7.2. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG, λ
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry