Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD48/SLAMF2, Clone: HM48-1, Novus Biologicals™

$225.00 - $397.00

Specifications

Antigen CD48/SLAMF2
Clone HM48-1
Immunogen The immunogen for this antibody was CD48.
Gene ID (Entrez) 962
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14340401

 
novus biologicals canada
NBP14340401MG
0.1mg Each for $278.00

NBP14340405

 
novus biologicals canada
NBP143404005MG
0.05mg Each for $225.00

NBP1434045

 
novus biologicals canada
NBP14340405MG
0.5mg Each for $397.00
Description & Specifications

Specifications

Antigen CD48/SLAMF2
Clone HM48-1
Immunogen The immunogen for this antibody was CD48.
Gene ID (Entrez) 962
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting