Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD48/SLAMF2, Clone: HM48-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143404005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14340405

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Mouse
Antigen CD48/SLAMF2
Host Species Armenian Hamster
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 962
Clone HM48-1
Formulation Phosphate buffer pH 7.2
Conjugate Unlabeled
Format Affinity Purified
Immunogen The immunogen for this antibody was CD48.
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.