Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD48/SLAMF2, Clone: HM48-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14340401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14340401

  • / Each
 

Description & Specifications

Specifications

Antigen CD48/SLAMF2
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Quantity 0.1mg
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Formulation Phosphate buffer pH 7.2
Gene ID (Entrez) 962
Primary or Secondary Primary
Format Affinity Purified
Immunogen The immunogen for this antibody was CD48.
Clone HM48-1
Regulatory Status RUO
Isotype IgG
Purification Method Affinity Purified
Host Species Armenian Hamster