Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD30/TNFRSF8, Clone: mCD30.1, Biotin, Novus Biologicals™

$175.00 - $582.00

Specifications

Antigen CD30/TNFRSF8
Clone mCD30.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD30.
Gene ID (Entrez) 943
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14365601

 
novus biologicals canada
NBP14365601MG
0.1mg Each for $252.00

NBP14365605

 
novus biologicals canada
NBP143656005MG
0.05mg Each for $175.00

NBP1436565

 
novus biologicals canada
NBP14365605MG
0.5mg Each for $582.00
Description & Specifications

Specifications

Antigen CD30/TNFRSF8
Clone mCD30.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD30.
Gene ID (Entrez) 943
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry