Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD27/TNFRSF7, Clone: LG.3A10, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD27/TNFRSF7
Clone LG.3A10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD27.
Gene Accession No. P26842
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14342701

 
novus biologicals canada
NBP14342701MG
0.1mg Each for $285.00

NBP14342703

 
novus biologicals canada
NBP1434270025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen CD27/TNFRSF7
Clone LG.3A10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD27.
Gene Accession No. P26842
Gene ID (Entrez) 939
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation