Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD27/TNFRSF7, Clone: LG.3A10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434270025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14342703

  • / Each
 

Description & Specifications

Specifications

Antigen CD27/TNFRSF7
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Clone LG.3A10
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Gene Accession No. P26842
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD27.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 939
Target Species Human
Target Species Murine