Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD27/TNFRSF7, Clone: LG.3A10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14342701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14342701

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Antigen CD27/TNFRSF7
Format Affinity Purified
Formulation PBS [pH 7.2]
Gene Accession No. P26842
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD27.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.1mg
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 939
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Clone LG.3A10
Concentration 0.5mg/mL