Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD11c, Clone: N418, PE, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen CD11c
Clone N418
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was PE CD11c.
Gene ID (Entrez) 3687
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14389001

 
novus biologicals canada
NBP14389001MG
0.1mg Each for $252.00

NBP14389002

 
novus biologicals canada
NBP14389002MG
0.2mg Each for $395.00

NBP14389005

 
novus biologicals canada
NBP143890005MG
0.05mg Each for $197.00
Description & Specifications

Specifications

Antigen CD11c
Clone N418
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was PE CD11c.
Gene ID (Entrez) 3687
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry