Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD11c, Clone: N418, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14344105MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1434415

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD11c.
Isotype IgG
Quantity 0.5mg
Clone N418
Conjugate Unlabeled
Cross Reactivity Mouse
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3687
Antigen CD11c
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL