Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD11c, Clone: N418, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143441005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344105

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Format Affinity Purified
Immunogen The immunogen for this antibody was CD11c.
Conjugate Unlabeled
Antigen CD11c
Host Species Armenian Hamster
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.05mg
Formulation Phosphate buffer pH 7.2
Cross Reactivity Mouse
Purification Method Affinity Purified
Isotype IgG
Primary or Secondary Primary
Concentration 0.5mg/mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone N418
Gene ID (Entrez) 3687