Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD11c, Clone: N418, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14344101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344101

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Host Species Armenian Hamster
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Clone N418
Concentration 0.5mg/mL
Regulatory Status RUO
Cross Reactivity Mouse
Formulation Phosphate buffer pH 7.2
Isotype IgG
Antigen CD11c
Conjugate Unlabeled
Quantity 0.1mg
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Format Affinity Purified
Primary or Secondary Primary
Gene ID (Entrez) 3687
Immunogen The immunogen for this antibody was CD11c.