Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD11c, Clone: N418, Cy5.5, PerCP, Novus Biologicals™

$175.00 - $395.00

Specifications

Antigen CD11c
Clone N418
Immunogen The immunogen for this antibody was CD11c.
Gene ID (Entrez) 3687
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14285801

 
novus biologicals canada
NBP14285801MG
0.1mg Each for $395.00

NBP14285803

 
novus biologicals canada
NBP1428580025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen CD11c
Clone N418
Immunogen The immunogen for this antibody was CD11c.
Gene ID (Entrez) 3687
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Cy5.5, PerCP
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry