Please login to your online account to display your discounted pricing

Armenian Hamster anti-CD11c, Clone: N418, Biotin, Novus Biologicals™

$131.00 - $395.00

Specifications

Antigen CD11c
Clone N418
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD11c.
Gene ID (Entrez) 3687
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14367701

 
novus biologicals canada
NBP14367701MG
0.1mg Each for $197.00

NBP14367705

 
novus biologicals canada
NBP143677005MG
0.05mg Each for $131.00

NBP1436775

 
novus biologicals canada
NBP14367705MG
0.5mg Each for $395.00
Description & Specifications

Specifications

Antigen CD11c
Clone N418
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD11c.
Gene ID (Entrez) 3687
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry