Please login to your online account to display your discounted pricing

Armenian Hamster anti-CCL2/MCP1, Clone: 4E2, Novus Biologicals™

$274.00 - $362.00

Specifications

Antigen CCL2/MCP1
Clone 4E2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CCL2.
Gene ID (Entrez) 6347
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14324305

 
novus biologicals canada
NBP143243005MG
0.05mg Each for $274.00

NBP1432435

 
novus biologicals canada
NBP14324305MG
0.5mg Each for $362.00
Description & Specifications

Specifications

Antigen CCL2/MCP1
Clone 4E2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CCL2.
Gene ID (Entrez) 6347
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA