Please login to your online account to display your discounted pricing

Armenian Hamster anti-CCL2/MCP1, Clone: 4E2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143243005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14324305

  • / Each
 

Description & Specifications

Specifications

Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Applications ELISA
Immunogen The immunogen for this antibody was CCL2.
Isotype IgG
Regulatory Status RUO
Gene ID (Entrez) 6347
Clone 4E2
Cross Reactivity Mouse
Antigen CCL2/MCP1
Format Affinity Purified
Purification Method Affinity Purified
Quantity 0.05mg
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Conjugate Unlabeled