Please login to your online account to display your discounted pricing

Armenian Hamster anti-CCL2/MCP1, Clone: 2H5, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen CCL2/MCP1
Clone 2H5
Concentration 0.5mg/mL
Immunogen CHO-expressed
Gene ID (Entrez) 6347
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1434905

 
novus biologicals canada
NBP14349005MG
0.5mg Each for $725.00

NBP143495MM

 
novus biologicals canada
NBP143490005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen CCL2/MCP1
Clone 2H5
Concentration 0.5mg/mL
Immunogen CHO-expressed
Gene ID (Entrez) 6347
Formulation Phosphate buffer (pH 7.2) with 150mM NaCl. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Flow Cytometry