Please login to your online account to display your discounted pricing

Armenian Hamster anti-B7-1/CD80, Clone: 16-10A1, Biotin, Novus Biologicals™

$175.00 - $428.00

Specifications

Antigen B7-1/CD80
Clone 16-10A1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14361701

 
novus biologicals canada
NBP14361701MG
0.1mg Each for $197.00

NBP14361705

 
novus biologicals canada
NBP143617005MG
0.05mg Each for $175.00

NBP1436175

 
novus biologicals canada
NBP14361705MG
0.5mg Each for $428.00
Description & Specifications

Specifications

Antigen B7-1/CD80
Clone 16-10A1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry