Please login to your online account to display your discounted pricing

Armenian Hamster anti-B7-1/CD80, Clone: 16-10A1, APC, Novus Biologicals™

$197.00 - $329.00

Specifications

Antigen B7-1/CD80
Clone 16-10A1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14303005

 
novus biologicals canada
NBP143030005MG
0.05mg Each for $197.00

NBP14303001

 
novus biologicals canada
NBP14303001MG
0.1mg Each for $329.00
Description & Specifications

Specifications

Antigen B7-1/CD80
Clone 16-10A1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Isotype IgG
Conjugate Allophycocyanin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry