Please login to your online account to display your discounted pricing

Armenian Hamster anti-AMICA/JAML, Clone: 4E10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14330905MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433095

  • / Each
 

Description & Specifications

Specifications

Antigen AMICA/JAML
Applications Flow Cytometry
Applications Immunoprecipitation
Clone 4E10
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was JAML.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 120425
Target Species Murine