Thermo Scientific™ Armadillo™ 96-Well PCR Plate
Optimize robotic applications with these ultra-rigid 96-well PCR plates with U-bottom wells, available in various colors.
$387.45 - $507.27
Specifications
Volume (Metric) Well 0.2mL
Material Polycarbonate frame with polypropylene wells
View More Specs
Disclaimers:
This product is sold under license from Eppendorf AG.
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Color Well Color Price Quantity  
Catalog Number Mfr. No. Description Color Well Color Price Quantity  
AB2396 Thermo Scientific™
AB-2396
Without bar code Clear Clear Case of 25 Each for $387.45
AB2396B Thermo Scientific™
AB-2396/B
Without bar code Blue Clear Case of 25 Each for $387.45
AB2396G Thermo Scientific™
AB2396/G
Without bar code Green Clear Case of 25 Each for $387.45
AB2396O Thermo Scientific™
AB-2396/O
Without bar code Orange Clear Case of 25 Each for $387.45
AB2396R Thermo Scientific™
AB-2396/R
Without bar code Red Clear Case of 25 Each for $387.45
AB2396Y Thermo Scientific™
AB-2396/Y
Without bar code Yellow Clear Case of 25 Each for $387.45
AB3396 Thermo Scientific™
AB-3396
Without bar code Clear White Case of 25 Each for $425.91
AB3396B Thermo Scientific™
AB-3396/B
Without bar code Blue White Case of 25 Each for $425.91
AB3396G Thermo Scientific™
AB-3396/G
Without bar code Green White Case of 25 Each for $425.91
AB3396O Thermo Scientific™
AB-3396/O
Without bar code Orange White Case of 25 Each for $425.91
BC2396 Thermo Scientific™
BC-2396
With bar code Clear Clear Case of 25 Each for $462.33
BC2396B Thermo Scientific™
BC-2396/B
With bar code Blue Clear Case of 25 Each for $462.33
BC2396G Thermo Scientific™
BC-2396/G
With bar code Green Clear Case of 25 Each for $462.33
BC2396O Thermo Scientific™
BC-2396/O
With bar code Orange Clear Case of 25 Each for $462.33
BC2396R Thermo Scientific™
BC-2396/R
With bar code Red Clear Case of 25 Each for $462.33
BC2396Y Thermo Scientific™
BC-2396/Y
With bar code Yellow Clear Case of 25 Each for $462.33
BC3396 Thermo Scientific™
BC-3396
With bar code Clear White Case of 25 Each for $507.27
BC3396B Thermo Scientific™
BC-3396/B
With bar code Blue White Case of 25 Each for $507.27
BC3396G Thermo Scientific™
BC-3396/G
With bar code Green White Case of 25 Each for $507.27
BC3396O Thermo Scientific™
BC-3396/O
With bar code Orange White Case of 25 Each for $507.27
BC3396R Thermo Scientific™
BC-3396/R
With bar code Red White Case of 25 Each for $507.27
BC3396Y Thermo Scientific™
BC-3396/Y
With bar code Yellow White Case of 25 Each for $507.27
AB3396R Thermo Scientific
AB-3396/R
Without bar code Red White Case of 25 Each for $425.94
AB3396Y Thermo Scientific
AB-3396/Y
Without bar code Yellow White Case of 25 Each for $425.94
Specifications
Description
Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping.
Highlights
  • Optically clear skirt - Aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • Raised rim design - Improved well surface area improves seal integrity in automated heat sealing
  • ANSI:SBS footprint - With rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high throughput applications
  • Thermostable polycarbonate frame design - Warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
  • Thin walled U-bottom wells - Optimal heat transfer and maximum sample recovery
  • Indented Alphanumeric Grid reference lettering - Integrated frame and well design for improved heat sealing
  • Low profile design - Allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (200 µL working volume)
  • Available in both clear and white coloured wells - Ensures the highest level of sensitivity for all PCR/qPCR reactions
  • Barcoding available - All plates can be provided with standard barcoding or barcoded to customer specifications for fully automated applications
Description & Specifications
Specifications
0.2mL
Polycarbonate frame with polypropylene wells