Please login to your online account to display your discounted pricing
Thermo Scientific™ Armadillo™ 96-Well PCR Plate
Optimize robotic applications with these ultra-rigid 96-well PCR plates with U-bottom wells, available in various colors.
Manufacturer: thermo scientific™  BC-3396/Y
 View more versions of this product
 Disclaimers
This product is sold under license from Eppendorf AG.
Catalog No. BC3396Y
  • / Case of 25 Each
Description & Specifications
Specifications
Description With bar code
Material Polycarbonate frame with polypropylene wells
Skirt Color Yellow
Volume (Metric) Well 0.2mL
Well Color White
Color Yellow
Item Description Yellow skirt; White well; Barcoded
Material Polycarbonate frame with polypropylene wells
Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping.
Armadillo plates are available in 96- and 384-well formats, multiple colors, and they can be ordered with a standard 128 barcode or custom barcoding. All formats are available with clear or white wells; choose white for superior qPCR results. The specially designed warp-resistant frame and multiple format options make Armadillo PCR plates the ultimate choice for high-throughput PCR and automated handling.
  • Polycarbonate frame for warp-resistant thermal cycling
  • Enhanced mechanical stability for robotic handling
  • Thin-walled polypropylene wells for optimal heat transfer
  • Well shape optimized for maximum sample recovery
  • Flat alpha-numeric lettering and raised rim well design for improved heat sealing
  • Optically-clear deck allows for easy visibility of wells
  • Multiple frame color options, all available in both clear (for PCR) and white colored wells (for qPCR)
  • Maximum well volume 0.2 mL