Please login to your online account to display your discounted pricing
Thermo Scientific™ Armadillo™ 96-Well PCR Plate
Optimize robotic applications with these ultra-rigid 96-well PCR plates with U-bottom wells, available in various colors.
Manufacturer: thermo scientific™  BC-3396/R
 View more versions of this product
 Disclaimers
This product is sold under license from Eppendorf AG.
Catalog No. BC3396R
  • / Case of 25 Each
Description & Specifications
Specifications
Color Red
Material Polycarbonate frame with polypropylene wells
Volume (Metric) Well 0.2mL
Skirt Color Red
Well Color White
Description With bar code
Material Polycarbonate frame with polypropylene wells
Item Description Red skirt; White well; Barcoded
Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping.
Armadillo plates are available in 96- and 384-well formats, multiple colors, and they can be ordered with a standard 128 barcode or custom barcoding. All formats are available with clear or white wells; choose white for superior qPCR results. The specially designed warp-resistant frame and multiple format options make Armadillo PCR plates the ultimate choice for high-throughput PCR and automated handling.
  • Polycarbonate frame for warp-resistant thermal cycling
  • Enhanced mechanical stability for robotic handling
  • Thin-walled polypropylene wells for optimal heat transfer
  • Well shape optimized for maximum sample recovery
  • Flat alpha-numeric lettering and raised rim well design for improved heat sealing
  • Optically-clear deck allows for easy visibility of wells
  • Multiple frame color options, all available in both clear (for PCR) and white colored wells (for qPCR)
  • Maximum well volume 0.2 mL