Please login to your online account to display your discounted pricing
Thermo Scientific™ Armadillo™ 384-Well PCR Plates
Optimize high throughput robotic PCR and qPCR applications with these uItra-rigid 384-well PCR plates with poylcarbonate frames and polypropylene wells.
$677.21 - $927.82
Specifications
Volume (Metric) Well 40µL
Material Polycarbonate frame with polypropylene wells
Volume (Metric) Working 25µL
View More Specs
 Disclaimers
This product is sold under license from Eppendorf AG.
Products
Catalog Number Mfr. No. Description Color Well Color Price Quantity    
AB2384
thermo scientific™
AB-2384
Without bar code Clear Clear Case of 50 Each for $677.21
AB2384B
thermo scientific™
AB-2384/B
Without bar code Blue Clear Case of 50 Each for $677.21
AB2384G
thermo scientific™
AB-2384/G
Without bar code Green Clear Case of 50 Each for $677.21
AB2384O
thermo scientific™
AB-2384/O
Without bar code Orange Clear Case of 50 Each for $677.21
AB2384R
thermo scientific™
AB-2384/R
Without bar code Red Clear Case of 50 Each for $677.21
AB2384Y
thermo scientific™
AB-2384/Y
Without bar code Yellow Clear Case of 50 Each for $677.21
AB3384
thermo scientific™
AB-3384
Without bar code Clear White Case of 50 Each for $746.15
AB3384B
thermo scientific™
AB-3384/B
Without bar code Blue White Case of 50 Each for $746.15
AB3384G
thermo scientific™
AB-3384/G
Without bar code Green White Case of 50 Each for $746.15
AB3384O
thermo scientific™
AB-3384/O
Without bar code Orange White Case of 50 Each for $746.15
AB3384R
thermo scientific™
AB-3384/R
Without bar code Red White Case of 50 Each for $746.15
AB3384Y
thermo scientific™
AB-3384/Y
Without bar code Yellow White Case of 50 Each for $746.15
BC2384
thermo scientific™
BC-2384
With bar code Clear Clear Case of 50 Each for $843.03
BC2384R
thermo scientific™
BC-2384/R
With bar code Red Clear Case of 50 Each for $843.03
BC3384
thermo scientific™
BC-3384
With bar code Clear White Case of 50 Each for $927.82
BC3384Y
thermo scientific™
BC-3384/Y
With bar code Yellow White Case of 50 Each for $927.82
BC2384B
thermo scientific
BC-2384/B
With bar code Blue Clear Case of 25 Each for $843.06
BC2384G
thermo scientific
BC-2384/G
With bar code Green Clear Case of 25 Each for $843.06
BC2384O
thermo scientific
BC-2384/O
With bar code Orange Clear Case of 25 Each for $843.06
BC2384Y
thermo scientific
BC-2384/Y
With bar code Yellow Clear Case of 25 Each for $843.06
BC3384G
thermo scientific
BC-3384/G
With bar code Green White Case of 25 Each for $927.82
BC3384B
thermo scientific
BC-3384/B
With bar code Blue White Case of 25 Each for $927.82
BC3384R
thermo scientific
BC-3384/R
With bar code Red White Case of 25 Each for $927.82
BC3384O
thermo scientific
BC-3384/O
With bar code Orange White Case of 25 Each for $927.82
Description & Specifications
Specifications
Volume (Metric) Well 40µL
Material Polycarbonate frame with polypropylene wells
Volume (Metric) Working 25µL
Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping.
The Armadillo PCR plates are available with clear or white wells for easy visibility of wells, and they feature thin-walled wells for optimal heat transfer.
Highlights
  • Optically clear skirt - Aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • Raised rim design - Improved well surface area improves seal integrity in automated heat sealing
  • ANSI:SBS footprint - With rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
  • Thermostable polycarbonate frame design - Warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
  • Thin walled U-bottom wells - Optimal heat transfer and maximum sample recovery
  • Indented Alphanumeric Grid reference lettering - Integrated frame and well design for improved heat sealing
  • Low profile design - Allows for maximum heat transfer and minimal evaporation when utilizing low-volume samples (30 µL working volume)
  • Available in both clear and white colored wells - Ensures the highest level of sensitivity for all PCR/qPCR reactions
  • Barcoding available - All plates are available already barcoded and can be provided barcoded to customer specifications for fully-automated applications