missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aristolochic acid, 96%, mixture of I and II, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC226095000

 View more versions of this product

Catalog No. AC226095000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Aristolochic acid
313-67-7
269°C to 270°C
Authentic
MFCD00004996
Glass bottle
aristolochiazaeure, aristolochic acid, aristolochic acid a, aristolochic acid i, aristolochic acid-i, aristolochin, aristolochine, birthwort, tardolyt, unii-94218wfp5t
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform,ether,acetone,,acetic acid,aniline,alkalies,practically insoluble in benzene,carbon disulfide
COC1=CC=CC2=C3C(=C(C=C21)[N+](=O)[O-])C(=CC4=C3OCO4)C(=O)O
2236
341.27
96%
Mixture of I and II
Yellow
95% min. (typically I+ II isomers) (HPLC)
C17H11NO7
19, V,7, 611
15, 777
500mg
BBFQZRXNYIEMAW-UHFFFAOYSA-N
341.27
CHEBI:2825
Powder

Chemical Identifiers

313-67-7
341.27
BBFQZRXNYIEMAW-UHFFFAOYSA-N
2236
COC1=CC=CC2=C3C(=C(C=C21)[N+](=O)[O-])C(=CC4=C3OCO4)C(=O)O
C17H11NO7
MFCD00004996
aristolochiazaeure, aristolochic acid, aristolochic acid a, aristolochic acid i, aristolochic acid-i, aristolochin, aristolochine, birthwort, tardolyt, unii-94218wfp5t
CHEBI:2825
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
Wear protective gloves/protective clothing/e

GHS Signal Word: Danger

EINECSNumber : 206-238-3

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only