Learn More
Aristolochic acid, 96%, mixture of I and II, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 226091000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aristolochic acid | |
313-67-7 | |
Yellow | |
95% min. (typically I+ II isomers) (HPLC) | |
C17H11NO7 | |
100 mg | |
15, 777 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform,ether,acetone,,acetic acid,aniline,alkalies,practically insoluble in benzene,carbon disulfide | |
COC1=CC=CC2=C3C(=C(C=C21)[N+](=O)[O-])C(=CC4=C3OCO4)C(=O)O | |
2236 | |
341.27 | |
Powder |
Mixture of I and II | |
269°C to 270°C | |
Authentic | |
Glass bottle | |
MFCD00004996 | |
19, V,7, 611 | |
aristolochic acid, aristolochic acid a, aristolochic acid i, aristolochin, tardolyt, birthwort, aristolochic acid-i, aristolochine, aristolochiazaeure, unii-94218wfp5t | |
BBFQZRXNYIEMAW-UHFFFAOYSA-N | |
341.27 | |
CHEBI:2825 | |
96% |
Chemical Identifiers
313-67-7 | |
341.27 | |
BBFQZRXNYIEMAW-UHFFFAOYSA-N | |
2236 | |
COC1=CC=CC2=C3C(=C(C=C21)[N+](=O)[O-])C(=CC4=C3OCO4)C(=O)O |
C17H11NO7 | |
MFCD00004996 | |
aristolochic acid, aristolochic acid a, aristolochic acid i, aristolochin, tardolyt, birthwort, aristolochic acid-i, aristolochine, aristolochiazaeure, unii-94218wfp5t | |
CHEBI:2825 |
Safety and Handling
GHS H Statement
May cause cancer.
Toxic if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger
EINECSNumber : 206-238-3
RUO – Research Use Only