Please login to your online account to display your discounted pricing
Argos Technologies HandE-Vac™ Handheld Aspirator
Rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes
$536.92 - $536.92
Products
Catalog Number Mfr. No. Price Quantity    
10-987-035
argos technologies
EV514
Each for $536.92
Description & Specifications
  • Ergonomic, handheld aspirator
  • Aspirates a 96-well plate in 8 seconds
  • Can be used with Pasteur pipets, plastic aspiration pipets or a variety of adapters for specific applications
  • Pressure-sensitive button on top of unit controls specific amount of vacuum applied
  • Prevents accidental aspiration of cells or supernatant associated with normal vacuum techniques
  • Use with standard in-house vacuum source or with M-Vac or E-Vac aspiration systems
  • Autoclavable