Please login to your online account to display your discounted pricing

Argos Technologies Flexirack Pipetting Workstation


Versatile polypropylene rack functions as a pipetting workstation

Manufacturer: argos technologies  R6006

 View more versions of this product

Catalog No. 03-395-206

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Material Polypropylene
Holds 15mL Tubes
Product Type Tube Insert for 4 x 15mL tubes (Blue)
Positions 4.0
For Use With (Equipment) FlexiRack pipetting workstation
Quantity 5/pack
Autoclavable No
Color Blue