Please login to your online account to display your discounted pricing

Argos Technologies Flexirack Pipetting Workstation


Versatile polypropylene rack functions as a pipetting workstation

Manufacturer: argos technologies  R6005

 View more versions of this product

Catalog No. 03-395-205

  • / Pack of 5 Each
 

Description & Specifications

Specifications

For Use With (Equipment) FlexiRack pipetting workstation
Autoclavable No
Quantity 5/pack
Holds 5mL Tubes
Color Green
Autoclavable No
Positions 7.0
Material Polypropylene
Positions 7.0
Product Type Tube Insert for 7 x 5mL tubes (Green)
Color Green
Tube Size 5mL