Please login to your online account to display your discounted pricing

Argos Technologies Flexirack Pipetting Workstation


Versatile polypropylene rack functions as a pipetting workstation

Manufacturer: argos technologies  R6004

 View more versions of this product

Catalog No. 03-395-204

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Holds 1.5mL Tubes
Quantity 5/pack
Material Polypropylene
For Use With (Equipment) FlexiRack pipetting workstation
Color Purple
Autoclavable No
Positions 12.0
Product Type Tube Insert for 12 x 1.5mL tubes (Purple)