Please login to your online account to display your discounted pricing

Argos Technologies Black Microcentrifuge Tubes


Ideal for use with fluorescent probes or ethidium bromide

Manufacturer: argos technologies  T7100BK

 View more versions of this product

Catalog No. 03-391-161

  • / Pack of 500 Each
 

Description & Specifications

Specifications

Color Black
For Use With (Application) Ideal for use with fluorescent probes or ethidium bromide
Certifications/Compliance RNase, DNase-free
Capacity (Metric) 1.5mL
Autoclavable Yes
Cap Style Snap Cap
Max. Force 15000xG
Material Polypropylene