Please login to your online account to display your discounted pricing
Argos Technologies Accessories for Nova Microincinerator
Manufacturer: argos technologies  S3012
 View more versions of this product
Catalog No. 03-391-445
  • / Each
Description & Specifications
Specifications
Temperature (English) 1500°F
Temperature (Metric) 815°C
For Use With (Application) Heat nova micro incinerator, dries slides, sputums, exudates and other thick mucous fluids