Please login to your online account to display your discounted pricing
Argos Technologies Accessories for Nova Microincinerator
$147.51 - $638.54
Products
Catalog Number Mfr. No. Price Quantity    
03-391-444
argos technologies
S3013
Each for $147.51
03-391-445
argos technologies
S3012
Each for $158.46
03-391-192
argos technologies
S3011H
Each for $635.09
03-391-193
argos technologies
S3022H
Each for $638.54
Description & Specifications
Aluminum Three-Place Slide Dryer
  • Safely dries a variety of samples using the heat from your incinerator
  • Dries slides, sputums, exudates, and other thick mucous fluids
  • No morphological distortions
  • Easily attaches to the top of the burner face plate
  • Also dissolves epithelium using the KOH mycology technique
Aluminum Loop Rest
  • Protects loop handles from overheating or melting
  • Fits any loop
  • Attaches easily to all incinerators