Please login to your online account to display your discounted pricing

Argos Technologies 5mL Microcentrifuge Tubes


Ideal alternative to 15mL conical tubes when using volumes under 5mL

Manufacturer: argos technologies  T2079A

 View more versions of this product

Catalog No. 03-391-273

  • / Pack of 200 Each
 

Description & Specifications

Specifications

Cap Style Easy-to-open cap
Material Polypropylene
Label Type Labeling Area
Capacity (Metric) 5mL
Max. Force 14000xG
Color Green
Autoclavable Yes
For Use With (Application) Sample tagging and identification
Sterile No
Shape Conical
Certifications/Compliance ISO Class 8, RNase, DNase, pyrogen-free