missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Argos Technologies 5mL Microcentrifuge Tubes

Ideal alternative to 15mL conical tubes when using volumes under 5mL

Manufacturer:  Argos Technologies T2078A

 View more versions of this product

Catalog No. 03-391-272


 

Specifications Description & Specifications

Specifications

Easy-to-open cap
Polypropylene
5mL
No
Sample tagging and identification
Labeling Area
Yes
Conical
Yellow
14000xG
ISO Class 8, RNase, DNase, pyrogen-free