Please login to your online account to display your discounted pricing

Arginase Mouse anti-Human, Clone: sl6arg, eBioscience™

Mouse Monoclonal Antibody

$131.95 - $323.35

Specifications

Antigen Arginase
Clone sl6arg
Host Species Murine
Species Reactivity Human
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122867

 
life technologies
LS14977980
25μg Each for $131.95

501128597

 
life technologies
LS14977982
100μg Each for $323.35
Description & Specifications

Specifications

Antigen Arginase
Clone sl6arg
Host Species Murine
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody sl6arg recognizes human Arginase-1, a 35kDa enzyme that that helps eliminate nitrogen by converting L-arginine to urea and L-ornithine which further converts to polyamine. These polyamines are important for cell proliferation and removal of toxins that arise from protein degradation. Expression is found in the liver, neutrophils, as well as myeloid derived suppressor cells (MDSC) and neural stem cells. Expression in macrophages has not been determined in humans, while in mice expression in M2 macrophages is well established. Arginase-1 may be expressed in the myeloid population in breast cancer tumors and is typically found in the majority of hepatocellular carcinomas.