missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arginase-1 Mouse anti-Human, eFluor 660, Clone: sl6arg, eBioscience™

Mouse Monoclonal Antibody

$175.45 - $442.25

Specifications

Antigen Arginase-1
Clone sl6arg
Host Species Murine
Species Reactivity Human
Applications Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Microscopy
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124545 Life Technologies
LS50977980
25μg Each for $175.45
501124546 Life Technologies
LS50977982
100μg Each for $442.25
Specifications

Description

The monoclonal antibody sl6arg recognizes human Arginase-1, a 35kDa enzyme that that helps eliminate nitrogen by converting L-arginine to urea and L-ornithine which further converts to polyamine. These polyamines are important for cell proliferation and removal of toxins that arise from protein degradation. Expression is found in the liver, neutrophils, as well as myeloid derived suppressor cells (MDSC) and neural stem cells. Expression in macrophages has not been determined in humans, while in mice expression in M2 macrophages is well established. Arginase-1 may be expressed in the myeloid population in breast cancer tumors and is typically found in the majority of hepatocellular carcinomas.
Description & Specifications

Specifications

Arginase-1
Murine
Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Microscopy
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
sl6arg
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer