Please login to your online account to display your discounted pricing

Argatroban, Tocris

Potent thrombin inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Argatroban
Assay Percent Range >99%
Molecular Formula C23H36N6O5S
Formula Weight 508.63
Synonym (2R,4R)-1-[(2S)-5-[(aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulfonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

163710

 
r & d systems
RDS163710
10mg Each for $201.36

163750

 
r & d systems
RDS163750
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 508.63
Quantity 10mg
Chemical Name or Material Argatroban
Molecular Formula C23H36N6O5S
Assay Percent Range >99%
CAS 74863-84-6
Synonym (2R,4R)-1-[(2S)-5-[(aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulfonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C