Please login to your online account to display your discounted pricing

Alfa Aesar™ [Arg&superscript_0;] Met-Enkephalin

For cellular and molecular biology applications

Specifications

Product Type [Arg0] Met-Enkephalin
Molecular Weight 729.84
Storage Requirements -30° to -10°C
Sequence Arg-Tyr-Gly-Gly-Phe-Met
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66105LB0

 
alfa aesar™
J66105LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66105MCR

 
alfa aesar™
J66105MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Arg0] Met-Enkephalin
Molecular Weight 729.84
Storage Requirements -30° to -10°C
Sequence Arg-Tyr-Gly-Gly-Phe-Met
Format Solid
Regulatory Status RUO
Purification Method HPLC