Please login to your online account to display your discounted pricing

Alfa Aesar™ Arg-Lys-Arg-Ala-Arg-Lys-Glu


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66858LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66858LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 5mg
Product Type Arg-Lys-Arg-Ala-Arg-Lys-Glu
Format Solid
Purification Method HPLC
Storage Requirements -30° to -10°C
Molecular Weight 943.12
Regulatory Status RUO